siRNA Details
| Gene Symbol: Tmem88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128155.1 | NP_001121627.1 | 2 | 321 | ||
| GenBank mRNA | EF532356.1 | - | 2 | 211 | ||
| EF532357.1 | - | 2 | 203 | |||
| EF532358.1 | - | 2 | 203 | |||
| EF532359.1 | - | 2 | 211 | |||
| EF532360.1 | - | 2 | 203 | |||

Tmem88
Genomic Map