siRNA Details
| Gene Symbol: LOC497938 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017475.1 | NP_001017475.1 | 2, 3 | 227 | ||
| XM_008767853.1 | XP_008766075.1 | 1, 2 | 256 | |||
| XM_008767854.1 | XP_008766076.1 | 1, 2 | 256 | |||
| GenBank mRNA | BC079298.1 | - | 2, 3 | 227 | 7134515 | |

LOC497938
Genomic Map