siRNA Details
| Gene Symbol: LOC497940 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017476.1 | NP_001017476.1 | 1 | 465 | ||
| XM_006246777.2 | XP_006246839.1 | 1 | 612 | |||
| XM_006246778.2 | XP_006246840.1 | 1 | 616 | |||
| XM_006246781.2 | XP_006246843.1 | 1 | 613 | |||
| XM_006246782.2 | XP_006246844.1 | 1 | 609 | |||
| GenBank mRNA | BC079326.1 | - | 1 | 465 | 7135484 | |

LOC497940
Genomic Map