siRNA Details
| Gene Symbol: Gps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017477.1 | NP_001017477.1 | 9, 10 | 1174 | ||
| XM_006246785.1 | XP_006246847.1 | 10, 11 | 1124 | |||
| XM_008767856.1 | XP_008766078.1 | 9, 10 | 1199 | |||
| GenBank mRNA | BC083699.1 | - | 9, 10 | 1174 | 7190067 | |
| FQ233878.1 | - | 10, 11 | 1008 | |||

Gps2
Genomic Map