siRNA Details
| Gene Symbol: Cxcl16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017478.1 | NP_001017478.1 | 2 | 428 | ||
| GenBank mRNA | BC079312.1 | - | 2 | 428 | 7134808 | |
| DQ025528.1 | - | 2 | 704 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cxcl16| Gene Symbol: Cxcl16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017478.1 | NP_001017478.1 | 2 | 428 | ||
| GenBank mRNA | BC079312.1 | - | 2 | 428 | 7134808 | |
| DQ025528.1 | - | 2 | 704 | |||