siRNA Details
| Gene Symbol: RGD1565611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110155.1 | NP_001103625.1 | 2 | 749 | ||
| XM_006246910.2 | XP_006246972.1 | 2 | 748 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1565611| Gene Symbol: RGD1565611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110155.1 | NP_001103625.1 | 2 | 749 | ||
| XM_006246910.2 | XP_006246972.1 | 2 | 748 | |||