siRNA Details
| Gene Symbol: Proca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025758.1 | NP_001020929.1 | 4 | 937 | ||
| XM_008768093.1 | XP_008766315.1 | 4 | 843 | |||
| GenBank mRNA | BC098036.1 | - | 4 | 937 | 7456533 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Proca1| Gene Symbol: Proca1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025758.1 | NP_001020929.1 | 4 | 937 | ||
| XM_008768093.1 | XP_008766315.1 | 4 | 843 | |||
| GenBank mRNA | BC098036.1 | - | 4 | 937 | 7456533 | |