siRNA Details
| Gene Symbol: Heatr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247018.2 | XP_006247080.1 | 4 | 1428 | ||
| XM_006247020.2 | XP_006247082.1 | 4 | 1428 | |||
| XM_006247021.2 | XP_006247083.1 | 3 | 1243 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Heatr9| Gene Symbol: Heatr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247018.2 | XP_006247080.1 | 4 | 1428 | ||
| XM_006247020.2 | XP_006247082.1 | 4 | 1428 | |||
| XM_006247021.2 | XP_006247083.1 | 3 | 1243 | |||