siRNA Details
| Gene Symbol: Heatr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079897.1 | NP_001073366.1 | 10 | 1824 | ||
| GenBank mRNA | EF122002.1 | - | 10 | 1824 | ||
| EF122003.1 | - | 10 | 1824 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Heatr6| Gene Symbol: Heatr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079897.1 | NP_001073366.1 | 10 | 1824 | ||
| GenBank mRNA | EF122002.1 | - | 10 | 1824 | ||
| EF122003.1 | - | 10 | 1824 | |||