siRNA Details
| Gene Symbol: Rad51c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129777.1 | NP_001123249.1 | 7 | 932 | ||
| XM_006247106.1 | XP_006247168.1 | 7 | 1010 | |||
| GenBank mRNA | BC166862.1 | - | 7 | 839 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rad51c| Gene Symbol: Rad51c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001129777.1 | NP_001123249.1 | 7 | 932 | ||
| XM_006247106.1 | XP_006247168.1 | 7 | 1010 | |||
| GenBank mRNA | BC166862.1 | - | 7 | 839 | ||