siRNA Details
| Gene Symbol: Dgke | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039341.1 | NP_001034430.1 | 6, 7 | 1272 | ||
| XM_006247070.2 | XP_006247132.1 | 6, 7 | 1507 | |||
| XM_006247071.2 | XP_006247133.1 | 6, 7 | 1507 | |||
| GenBank mRNA | BC110049.1 | - | 6, 7 | 1272 | 7316890 | |

Dgke
Genomic Map