siRNA Details
| Gene Symbol: Med1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134361.1 | NP_001127833.1 | 17 | 4429 | ||
| XM_006247398.2 | XP_006247460.1 | 17 | 4579 | |||
| XM_006247399.2 | XP_006247461.1 | 18 | 4654 | |||
| XM_008768105.1 | XP_008766327.1 | 17 | 5209 | |||

Med1
Genomic Map