siRNA Details
Gene Symbol: Ccdc103 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109049.1 | NP_001102519.1 | 4 | 410 | ||
XM_006247538.2 | XP_006247600.1 | 3 | 264 | |||
XM_006247540.2 | XP_006247602.1 | 2 | 235 |
