siRNA Details
| Gene Symbol: Hexim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025136.1 | NP_001020307.1 | 1 | 1299 | ||
| GenBank mRNA | BC087133.1 | - | 1 | 1299 | 7122255 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hexim1| Gene Symbol: Hexim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025136.1 | NP_001020307.1 | 1 | 1299 | ||
| GenBank mRNA | BC087133.1 | - | 1 | 1299 | 7122255 | |