siRNA Details
| Gene Symbol: Ern1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191926.1 | NP_001178855.1 | 14 | 1708 | ||
| XM_006247633.2 | XP_006247695.1 | 14 | 1959 | |||
| XM_006247634.2 | XP_006247696.1 | 14 | 1952 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ern1| Gene Symbol: Ern1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191926.1 | NP_001178855.1 | 14 | 1708 | ||
| XM_006247633.2 | XP_006247695.1 | 14 | 1959 | |||
| XM_006247634.2 | XP_006247696.1 | 14 | 1952 | |||