siRNA Details
| Gene Symbol: Kctd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768411.1 | XP_008766633.1 | 5 | 715 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kctd2| Gene Symbol: Kctd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008768411.1 | XP_008766633.1 | 5 | 715 | ||