siRNA Details
| Gene Symbol: Fn3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109051.1 | NP_001102521.1 | 4 | 488 | ||
| XM_006247907.2 | XP_006247969.1 | 6 | 1384 | |||
| GenBank mRNA | FQ212712.1 | - | 8 | 949 | ||
| FQ212922.1 | - | 6 | 852 | |||

Fn3k
Genomic Map