siRNA Details
| Gene Symbol: Fn3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109051.1 | NP_001102521.1 | 4 | 585 | ||
| XM_006247907.2 | XP_006247969.1 | 6 | 1481 | |||
| GenBank mRNA | FQ212712.1 | - | 8 | 1046 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fn3k| Gene Symbol: Fn3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109051.1 | NP_001102521.1 | 4 | 585 | ||
| XM_006247907.2 | XP_006247969.1 | 6 | 1481 | |||
| GenBank mRNA | FQ212712.1 | - | 8 | 1046 | ||