siRNA Details
| Gene Symbol: LOC498063 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017483.1 | NP_001017483.1 | 1 | 566 | ||
| GenBank mRNA | BC083887.1 | - | 1 | 566 | 7132136 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC498063| Gene Symbol: LOC498063 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017483.1 | NP_001017483.1 | 1 | 566 | ||
| GenBank mRNA | BC083887.1 | - | 1 | 566 | 7132136 | |