siRNA Details
| Gene Symbol: Trat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037981.1 | NP_001033070.1 | 6 | 625 | ||
| GenBank mRNA | BC111081.1 | - | 6 | 625 | 7384735 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trat1| Gene Symbol: Trat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037981.1 | NP_001033070.1 | 6 | 625 | ||
| GenBank mRNA | BC111081.1 | - | 6 | 625 | 7384735 | |