siRNA Details
| Gene Symbol: Spidr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100988.1 | NP_001094458.1 | 12 | 1815 | ||
| XM_008768898.1 | XP_008767120.1 | 13 | 2011 | |||
| XM_008768899.1 | XP_008767121.1 | 13 | 2011 | |||
| GenBank mRNA | BC129120.1 | - | 12 | 1815 | 7123928 | |

Spidr
Genomic Map