siRNA Details
| Gene Symbol: Cdk8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109061.1 | NP_001102531.1 | 3 | 270 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdk8| Gene Symbol: Cdk8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109061.1 | NP_001102531.1 | 3 | 270 | ||