siRNA Details
| Gene Symbol: Spdye4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034153.1 | NP_001029325.1 | 1 | 213 | ||
| GenBank mRNA | BC105855.1 | - | 1 | 213 | 7453404 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spdye4| Gene Symbol: Spdye4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034153.1 | NP_001029325.1 | 1 | 213 | ||
| GenBank mRNA | BC105855.1 | - | 1 | 213 | 7453404 | |