siRNA Details
| Gene Symbol: Spdye4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034153.1 | NP_001029325.1 | 2 | 292 | ||
| XM_008768995.1 | XP_008767217.1 | 2 | 301 | |||
| XM_008768996.1 | XP_008767218.1 | 2 | 237 | |||
| GenBank mRNA | BC105855.1 | - | 2 | 292 | 7453404 | |

Spdye4
Genomic Map