siRNA Details
| Gene Symbol: LOC498155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047924.1 | NP_001041389.1 | 1, 2 | 70 | ||
| GenBank mRNA | AY539918.1 | - | 1, 2 | 70 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC498155| Gene Symbol: LOC498155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047924.1 | NP_001041389.1 | 1, 2 | 70 | ||
| GenBank mRNA | AY539918.1 | - | 1, 2 | 70 | ||