siRNA Details
| Gene Symbol: Spry3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109063.1 | NP_001102533.1 | 2 | 436 | ||
| XM_008769065.1 | XP_008767287.1 | 2 | 1810 | |||
| XM_008769066.1 | XP_008767288.1 | 3 | 731 | |||
| XM_008769067.1 | XP_008767289.1 | 3 | 683 | |||
| XM_008769068.1 | XP_008767290.1 | 2 | 399 | |||
| XM_008769069.1 | XP_008767291.1 | 2 | 529 | |||
| GenBank mRNA | BC168971.1 | - | 2 | 429 | 7316797 | |

Spry3
Genomic Map