siRNA Details
| Gene Symbol: Zkscan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025760.1 | NP_001020931.1 | 5, 6 | 988 | ||
| GenBank mRNA | BC099191.1 | - | 5, 6 | 988 | 7383463 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zkscan1| Gene Symbol: Zkscan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025760.1 | NP_001020931.1 | 5, 6 | 988 | ||
| GenBank mRNA | BC099191.1 | - | 5, 6 | 988 | 7383463 | |