siRNA Details
| Gene Symbol: Prkrip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098793.1 | NP_001092263.1 | 2, 3 | 236 | ||
| GenBank mRNA | BC158549.1 | - | 2, 3 | 236 | 8361725 | |
| DY314400.1 | - | 2, 3 | 236 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkrip1| Gene Symbol: Prkrip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098793.1 | NP_001092263.1 | 2, 3 | 236 | ||
| GenBank mRNA | BC158549.1 | - | 2, 3 | 236 | 8361725 | |
| DY314400.1 | - | 2, 3 | 236 | |||