siRNA Details
| Gene Symbol: Bri3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017487.1 | NP_001017487.1 | 3 | 551 | ||
| GenBank mRNA | BC085334.1 | - | 3 | 551 | 7301029 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bri3bp| Gene Symbol: Bri3bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017487.1 | NP_001017487.1 | 3 | 551 | ||
| GenBank mRNA | BC085334.1 | - | 3 | 551 | 7301029 | |