siRNA Details
| Gene Symbol: Mapkapk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025761.2 | NP_001020932.2 | 5 | 436 | ||
| NM_001164043.1 | NP_001157515.1 | 8 | 805 | |||
| GenBank mRNA | BC099161.1 | - | 5 | 436 | 7381631 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mapkapk5| Gene Symbol: Mapkapk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025761.2 | NP_001020932.2 | 5 | 436 | ||
| NM_001164043.1 | NP_001157515.1 | 8 | 805 | |||
| GenBank mRNA | BC099161.1 | - | 5 | 436 | 7381631 | |