siRNA Details
| Gene Symbol: RGD1562310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109066.1 | NP_001102536.1 | 1 | 178 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1562310| Gene Symbol: RGD1562310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109066.1 | NP_001102536.1 | 1 | 178 | ||