siRNA Details
| Gene Symbol: Ccdc60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034945.1 | NP_001030117.1 | 6 | 986 | ||
| XM_006249464.2 | XP_006249526.1 | 6 | 1003 | |||
| GenBank mRNA | BC103639.1 | - | 6 | 986 | 7133734 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc60| Gene Symbol: Ccdc60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034945.1 | NP_001030117.1 | 6 | 986 | ||
| XM_006249464.2 | XP_006249526.1 | 6 | 1003 | |||
| GenBank mRNA | BC103639.1 | - | 6 | 986 | 7133734 | |