siRNA Details
| Gene Symbol: Fam222a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109067.1 | NP_001102537.1 | 3 | 1711 | ||
| XM_008769352.1 | XP_008767574.1 | 2 | 4367 | |||
| XM_008769353.1 | XP_008767575.1 | 2 | 1316 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam222a| Gene Symbol: Fam222a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109067.1 | NP_001102537.1 | 3 | 1711 | ||
| XM_008769352.1 | XP_008767574.1 | 2 | 4367 | |||
| XM_008769353.1 | XP_008767575.1 | 2 | 1316 | |||