siRNA Details
| Gene Symbol: Mn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191928.1 | NP_001178857.1 | 1 | 90 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mn1| Gene Symbol: Mn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191928.1 | NP_001178857.1 | 1 | 90 | ||