siRNA Details
| Gene Symbol: LOC498222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100788.1 | NP_001094258.1 | 4 | 723 | ||
| XM_006249714.2 | XP_006249776.1 | 5 | 2985 | |||
| XM_008769488.1 | XP_008767710.1 | 4 | 848 | |||
| XM_008769489.1 | XP_008767711.1 | 3 | 2693 | |||

LOC498222
Genomic Map