siRNA Details
| Gene Symbol: Pm20d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109068.1 | NP_001102538.1 | 6 | 783 | ||
| GenBank mRNA | FQ210945.1 | - | 2 | 70 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pm20d1| Gene Symbol: Pm20d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109068.1 | NP_001102538.1 | 6 | 783 | ||
| GenBank mRNA | FQ210945.1 | - | 2 | 70 | ||