siRNA Details
| Gene Symbol: Mfsd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109069.1 | NP_001102539.1 | 6 | 1203 | ||
| XM_006249791.2 | XP_006249853.1 | 6 | 1198 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mfsd4| Gene Symbol: Mfsd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109069.1 | NP_001102539.1 | 6 | 1203 | ||
| XM_006249791.2 | XP_006249853.1 | 6 | 1198 | |||