siRNA Details
Gene Symbol: LOC498231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047925.1 | NP_001041390.1 | 6 | 592 | ||
GenBank mRNA | AY539895.1 | - | 6 | 592 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC498231 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047925.1 | NP_001041390.1 | 6 | 592 | ||
GenBank mRNA | AY539895.1 | - | 6 | 592 |