siRNA Details
| Gene Symbol: Lax1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017491.1 | NP_001017491.1 | 6 | 780 | ||
| XM_006249793.2 | XP_006249855.1 | 6 | 915 | |||
| GenBank mRNA | BC089837.1 | - | 6 | 780 | 7365271 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lax1| Gene Symbol: Lax1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017491.1 | NP_001017491.1 | 6 | 780 | ||
| XM_006249793.2 | XP_006249855.1 | 6 | 915 | |||
| GenBank mRNA | BC089837.1 | - | 6 | 780 | 7365271 | |