siRNA Details
| Gene Symbol: Apobec4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017492.1 | NP_001017492.1 | 2 | 853 | ||
| XM_008769666.1 | XP_008767888.1 | 2 | 1660 | |||
| XM_008769667.1 | XP_008767889.1 | 2 | 1064 | |||
| GenBank mRNA | BC079272.1 | - | 2 | 853 | 7134003 | |

Apobec4
Genomic Map