siRNA Details
| Gene Symbol: Cfap126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109075.1 | NP_001102545.1 | 2, 3 | 347 | ||
| XM_008769751.1 | XP_008767973.1 | 1, 2 | 954 | |||
| XM_008769752.1 | XP_008767974.1 | 1, 2 | 954 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfap126| Gene Symbol: Cfap126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109075.1 | NP_001102545.1 | 2, 3 | 347 | ||
| XM_008769751.1 | XP_008767973.1 | 1, 2 | 954 | |||
| XM_008769752.1 | XP_008767974.1 | 1, 2 | 954 | |||