siRNA Details
| Gene Symbol: Cfap126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109075.1 | NP_001102545.1 | 6 | 782 | ||
| XM_008769751.1 | XP_008767973.1 | 5 | 1389 | |||
| XM_008769752.1 | XP_008767974.1 | 4 | 1308 | |||
| GenBank mRNA | FQ219903.1 | - | 6 | 629 | ||

Cfap126
Genomic Map