siRNA Details
| Gene Symbol: Fcer1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131001.1 | NP_001124473.1 | 5 | 306 | ||
| GenBank mRNA | BC160864.1 | - | 5 | 306 | 7324585 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fcer1g| Gene Symbol: Fcer1g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131001.1 | NP_001124473.1 | 5 | 306 | ||
| GenBank mRNA | BC160864.1 | - | 5 | 306 | 7324585 | |