siRNA Details
| Gene Symbol: Arhgap30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109077.1 | NP_001102547.1 | 12 | 2898 | ||
| GenBank mRNA | FQ211641.1 | - | 2906 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Arhgap30| Gene Symbol: Arhgap30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109077.1 | NP_001102547.1 | 12 | 2898 | ||
| GenBank mRNA | FQ211641.1 | - | 2906 | |||