siRNA Details
| Gene Symbol: Itln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034946.1 | NP_001030118.1 | 8 | 931 | ||
| GenBank mRNA | BC099766.1 | - | 8 | 931 | 7931002 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Itln1| Gene Symbol: Itln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034946.1 | NP_001030118.1 | 8 | 931 | ||
| GenBank mRNA | BC099766.1 | - | 8 | 931 | 7931002 | |