siRNA Details
| Gene Symbol: Smyd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025762.1 | NP_001020933.1 | 254 | |||
| GenBank mRNA | BC097455.1 | - | 254 | 7457098 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smyd3| Gene Symbol: Smyd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025762.1 | NP_001020933.1 | 254 | |||
| GenBank mRNA | BC097455.1 | - | 254 | 7457098 | ||