siRNA Details
| Gene Symbol: Wdr26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109081.1 | NP_001102551.1 | 3, 4 | 337 | ||
| GenBank mRNA | FQ226610.1 | - | 331 | |||
| FQ227016.1 | - | 331 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr26| Gene Symbol: Wdr26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109081.1 | NP_001102551.1 | 3, 4 | 337 | ||
| GenBank mRNA | FQ226610.1 | - | 331 | |||
| FQ227016.1 | - | 331 | ||||