siRNA Details
| Gene Symbol: Vash2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109082.1 | NP_001102552.1 | 2, 3 | 479 | ||
| XM_006250463.2 | XP_006250525.1 | 2, 3 | 494 | |||
| XM_006250465.2 | XP_006250527.1 | 2, 3 | 494 | |||
| XM_008769868.1 | XP_008768090.1 | 2, 3 | 494 | |||
| XM_008769869.1 | XP_008768091.1 | 2, 3 | 817 | |||
| XM_008769870.1 | XP_008768092.1 | 2, 3 | 494 | |||
| XM_008769871.1 | XP_008768093.1 | 2, 3 | 494 | |||
| XM_008769872.1 | XP_008768094.1 | 2, 3 | 494 | |||
| GenBank mRNA | AY724511.1 | - | 2, 3 | 439 | ||

Vash2
Genomic Map