siRNA Details
| Gene Symbol: Nsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109083.1 | NP_001102553.1 | 6 | 791 | ||
| XM_006250488.1 | XP_006250550.1 | 6 | 787 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nsl1| Gene Symbol: Nsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109083.1 | NP_001102553.1 | 6 | 791 | ||
| XM_006250488.1 | XP_006250550.1 | 6 | 787 | |||