siRNA Details
| Gene Symbol: Coq2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044255.1 | NP_001037720.1 | 6, 7 | 946 | ||
| GenBank mRNA | BC099827.1 | - | 6, 7 | 946 | 7934175 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Coq2| Gene Symbol: Coq2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044255.1 | NP_001037720.1 | 6, 7 | 946 | ||
| GenBank mRNA | BC099827.1 | - | 6, 7 | 946 | 7934175 | |